Would you like to log in again?
SP Bel-Art Clavies™ Autoclavable Bags, Bel-Art Products, a part of SP
Supplier: Bel-Art
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
SP Bel-Art Clavies™
H13182-1012 H13185-2436 H13185-2430 H13185-1224
30617-610CS 2510.2 USD InStock
30617-610 30617-608 490005-658 490005-656
SP Bel-Art Clavies™ Autoclavable Bags, Bel-Art Products, a part of SP
Bags Waste Disposal Bags
Clavies® bags offer maximum protection and performance for autoclavibility.
  • Steam autoclavable at 135 °C (275 °F)
The durable 0.05 mm thick polypropylene bag is filled with used labware such as Petri dishes, membrane filters, culture plates, pipettes, and more, then autoclaved and discarded.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty