Would you like to log in again?
SP Bel-Art Poxygrid® Test Tube Racks, Bel-Art Products, a part of SP
Supplier: Bel-Art
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Poxygrid®
F18757-0000 187620000 F18773-0000 F18750-0000 F18764-0000 F18773-0001 F18750-0001 F18773-0002 F18757-0002 F18764-0001 F18755-0002 F18754-0000 F18772-0001 F18772-0002 F18759-0003 F18753-0003 F18754-0001 F18755-0001 F18757-0001 F18755-0003 F18772-0000_SE F18750-0002 F18753-0000 F18767-0000 F18762-0003 F18762-0002 F18773-0003 F18759-0000 F18772-0003 F18750-0003 187550000 F18762-0001 F18759-0002 F18757-0003 F18759-0001 F18767-0001
30617-630CS 1778.14 USD InStock
30617-630 30617-632 76326-290 76326-292 30617-638 76326-294 76326-330 76326-296 76326-332 76326-310 76326-298 30617-634 76326-334 76326-312 30617-636 76326-302 76326-324 76326-326 76326-328 76326-306 60917-715 76326-308 490005-578 30617-640 30617-642 80700-746 80700-748 30617-628 76326-288 76326-320 470145-332 76326-300 76326-322 76326-314 76326-316 76326-318
SP Bel-Art Poxygrid® Test Tube Racks, Bel-Art Products, a part of SP
Racks Tube Racks
Chemical- and corrosion-resistant steel wire racks are coated with heat-cured epoxy.
  • Epoxy coatings are unaffected by organic solvents, salts, weak acids, and weak alkalis
  • Slotted grid bottom and reinforced edges
  • Autoclavable
They will withstand a temperature range of -223 to 121 °C (-369 to 250 °F) and can be autoclaved.
All racks have square holes, three decks, a slotted grid bottom, and reinforced edges.
Orange racks can be used for biohazard warning, yellow for radioactive warning.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty