Would you like to log in again?
SP Bel-Art Spinpak® Octagon Magnetic Stirring Bar Packs, Bel-Art Products, a part of SP
Supplier: Bel-Art
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Spinpak®
F37157-0020 F37157-0005 F37157-0015 F37157-0010
76001-834CS 515.35 USD InStock
76001-834 30617-692 30617-694 30617-690
SP Bel-Art Spinpak® Octagon Magnetic Stirring Bar Packs, Bel-Art Products, a part of SP
Stirrers Magnetic Stirring Bars
Octagon magnetic stirring bars with integral pivot ring are the most commonly used shape
  • PTFE low-friction coating
  • Durable and inert
  • Interrupted profile provides greater surface area and added turbulence
  • Pivot ring aids in reducing friction and chattering
  • Autoclavable
Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification.
One size per pack.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty