Would you like to log in again?
SP Bel-Art Easy-Read Individually Calibrated Environmentally Friendly Liquid-In-Glass Thermometers, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a part of SP
B60202-0500 B60202-0100 B60202-0000 B60202-1200 B60202-1400 B60202-1300 30520C B60202-0300 30510C 30515C
89218-020EA 355.61 USD InStock
89218-020 89082-179 75795-782 75795-784 30617-808 30617-782 89218-024 75795-780 89218-028 75795-778
SP Bel-Art Easy-Read Individually Calibrated Environmentally Friendly Liquid-In-Glass Thermometers, Bel-Art Products, a part of SP
Thermometers Analog Thermometers
Thermometers contain black, non-toxic, biodegradable, EnviroKleen™ certified, Enviro-Safe® liquid against lead-free, yellow back glass for easy readability and fewer reading errors.
  • Non-toxic and biodegradable liquid
  • Lead-free
  • Yellow back glass
  • Easy to read
They are ideal for use in chemical/petrochemical laboratories, universities, scientific institutions, food and beverage processing, pharmaceutical and medical industries, dairies, breweries and distilleries. Individually serialized.
Ordering information: Each easy-to-read thermometer includes an individualized Thermometer Calibration Report stating corrections to one-tenth of the smallest division, unique report number and NIST standards used for the calibration.
Order Now
ORDER
SPECIFICATIONS
SERVICES
Services: VWRCATALYST™ offers convenient and efficient re-calibration services for your liquid in glass thermometers with an industry leading 5-day average turn-around time from receipt of your instrument. Simply order the stated catalog no. based on how many calibration points are desired. You will receive a protective box to ship your instrument to our certified calibration laboratory along with a work order form to specify your calibration points 0 to 100°C (32 to 212°F) and prepaid return shipping label. Upon receipt at the laboratory your instrument will be inspected for any defect under a microscope and the performance of your instrument will be compared to certified standards and corrections to one-tenth of the smallest division recorded on your signed calibration report. Your calibration report and instrument are carefully repacked then returned to you prepaid.
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty