Would you like to log in again?
THERMOMETER DIAL -10/100C
Supplier: Bel-Art Products, a Part of SP
THERMOMETER DIAL -10/100C^ DURAC* Bi-Metallic Dial Thermometers. Dial Size: 1.3in, Probe Length: 4.9in, Range: -10/110C
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
THERMOMETER DIAL -10/100C 30617-784 B61310-3400 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty