Would you like to log in again?
CELLSTAR® Cell-Repellent Surface Microplates and Dishes, Greiner Bio-One
Supplier: Greiner Bio-One
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
CELLSTAR®
781970 650979 781976 650970 657970 781976-SIN 651970 677970 655976-SIN 655970 655976 662970
10014-324CT 1125 USD InStock
10014-324 30618-022 10014-326 10014-316 10014-320 30618-026 76199-200 10014-322 10859-850 30618-028 10859-852 10014-318
CELLSTAR® Cell-Repellent Surface Microplates and Dishes, Greiner Bio-One
Microplates Cell Culture Plates
Greiner Bio-One introduces a new surface chemistry designed to effectively inhibit cell adhesion.
  • Nonpyrogenic
  • Free of RNase, DNase, and human DNA
  • Cell-repellent surface
  • Ideal substrate for native cell culture experiments
Achieved through an innovative chemical polymer modification, Greiner Bio-One’s cell-repellent surface does not degrade or leach under common cell culture conditions, rendering an ideal substrate for native cell culture experiments.
The cell-repellent surface is ideal for applications such as spheroid culture of tumor cells, spheroid culture of embryonic cells, embryoid body formation, and suspension culture of semi-adherent and adherent cell lines. All vessels are free of detectable RNase, DNase, and human DNA. nonpyrogenic and noncytotoxic.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty