Would you like to log in again?
HV Valve Fittings, Hamilton
Supplier: Hamilton
35031 88800 35073 35030 32834 32835 35071 32822 35070 32805 32823 86512 35033 35072 35032 32836 35037
65513-383EA 17.38 USD InStock
65513-383 65516-720 89221-480 65513-340 65513-420 65516-722 65513-146 65513-102 65516-719 89221-416 30618-690 89221-424 89221-426 65513-645 65513-601 65513-689 65513-306
HV Valve Fittings, Hamilton
Valves Vacuum Valves
Use the female luer, male luer, luer lock and barbed fittings to connect syringes, equipment, or other devices to Hamilton valves.
Hamilton HV, HVP, HVX, HVM, and HVXM valves have a port depth of 5.6 mm (7/32") which is much shorter than the industry standard ports
Use tubing assemblies to connect tubing to valves (100psig or less). Fittings and adapters are constructed of Kel-F® material (CTFE), except the male luer lock fittings and swivel union, which are made of Kel-F® (CTFE) and metal. Some fittings are offered in two port diameters for fluid flow optimization.
This design minimizes the dead volume that is trapped in the valve port. To take advantage of this unique feature, a line of short adapters has been designed that will only work with Hamilton valves. These fittings have a 1/4"-28 UNF thread connection on one side and either a male luer, female luer, or male luer lock on the other side."
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty