Would you like to log in again?
Laboratory Bottle, Narrow Mouth, HDPE, Bulk Packed, Dynalon
Supplier: Dynalon
301715-0008
30620-176CS 468.34 USD InStock
30620-176
Laboratory Bottle, Narrow Mouth, HDPE, Bulk Packed, Dynalon
Bottles Plastic Bottles
Leakproof, rigid, and translucent high density polyethylene narrow mouth lab bottles offer excellent chemical resistance.
  • Ideal for storage and handling of liquids or solids
Manufactured from FDA approved resin for non-contaminating storage of chemicals and food grade products. Bulk packed with closures included but not attached.
Delivery information: Bottles are supplied with polypropylene closures.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty