Would you like to log in again?
Laboratory Bottles, Wide Mouth, LDPE, Bulk Packed, Dynalon
Supplier: Dynalon
301675-0008 301675-0004 301675-0032 301675-0016
30620-184CS 700.17 USD InStock
30620-184 30620-182 30621-002 30620-186
Laboratory Bottles, Wide Mouth, LDPE, Bulk Packed, Dynalon
Bottles Plastic Bottles
Translucent, low density polyethylene wide mouth bottles are supplied with leakproof polypropylene screw closures.
Manufactured from FDA approved resin for use with food products. The LDPE construction offers greater flexibility and impact resistance than HDPE bottles. Bulk packed with closures included but not attached.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty