Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bottle, Wide Mouth, PTFE, Dynalon

Supplier: Dynalon
314874
30620-196EA 502.54 USD InStock
30620-196
Bottle, Wide Mouth, PTFE, Dynalon
Bottles Plastic Bottles
Heavy wall, wide mouth air tight PTFE bottles with screw closure have machine finish for ease in cleaning.

  • Working temperature range: Cryogenic to 260 °C
  • Non-sticking
  • Wall thickness: 3 to 4 mm

Virtually immune to chemical attack, only attacked by some fluorine containing compounds at high temperature and molten alkalis metals.

Ordering information: Elasticity of tubing will influence choice of size. Dimensions listed are nominal.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty