Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Wide Neck Wash Bottles, Color Coded, Dynalon
Supplier: Dynalon
506935-0004 506925-0001 506945-0003 506935-0005 506945-0001 506945-0004 506945-0005 506925-0002 506925-0003 506935-0002 506945-0002 506925-0004 506925-0005 506935-0001
30620-202CS 74.38 USD InStock
30620-202 30620-214 30620-236 30620-200 30620-212 30620-234 30620-220 30620-232 30620-210 30620-230 30620-218 30620-204 30620-238 30620-216
Wide Neck Wash Bottles, Color Coded, Dynalon
Bottles Wash Bottles
High-quality natural polyethylene wash bottles are fitted with a leak-proof, wide neck, color coded polypropylene spout closure.
The wide neck offers easy spill-free filling, and the acute angled, precision-molded dispensing spout ensures a consistent downward pointing jet. The wash bottles are supplied in shrink-wrapped packs of five. All vessels feature S.P.I recycling codes on the base. Maximum operating temperature of LDPE is 80 °C (176 °F).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty