Would you like to log in again?
Polypropylene Carboys, Dynalon
Supplier: Dynalon
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
505634-1 505634-3 505634-1 505634-2 505634-2 505634-3
30620-250EA 153.58 USD InStock
30620-250 470200-406 470200-408 30620-248 30620-246 470202-528
Polypropylene Carboys, Dynalon
Bottles Carboys
Rounded octagonal shape for increased rigidity when full.
  • Heavy-duty autoclavable polypropylene construction
  • Offset neck enables easier filling and pouring
  • Comfortable blow molded handles allow easy carrying
Heavy-duty closures are compatible with other leading brands. Carboys feature a durable and leak-proof heavy-duty stopcock with boss fitting for extra strength. Stopcock features removable tube fitting for added convenience. Graduations are molded into the front of the product, allowing easy reading even when several carboys are used in a row.
Caution: The silicone seal can be damaged by HCL.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty