Would you like to log in again?
Printed Cylinders, Dynalon
Supplier: Dynalon
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
537805-0100 537815-1000 537815-0500 537805-0010 537805-1000 537805-0250 537805-2000 537815-2000 537805-0050 537805-0025 537805-0500 537815-0050 537815-0100
30620-282CS 223.74 USD InStock
30620-282 30620-294 30620-280 30620-292 30620-290 30620-302 30620-300 30620-286 30620-298 30620-284 30620-296 30620-308 30620-306
Printed Cylinders, Dynalon
Cylinders Measuring Cylinders
  • Brilliant easy to read blue printed graduations
  • Shatter resistant - reduces breakage and injury
  • Chemically resistant to most acids, bases and many common solvents
  • Autoclavable
  • Choose glass-like clarity of polymethylpentene (PMP) or
  • Choose economical translucent polypropylene (PP)
  • Food grade product
These cylinders feature permanently easy to read blue screened graduations that resist fading and scratching after repeated autoclaving. Meets stringent international quality criteria according to ISO 6706 standards. Tolerances according to DIN 12 680 TT, Class B. Maximum temperature: PP 121°C, PMP 135°C.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty