Would you like to log in again?
Jar, Straight Sided, Polypropylene, Dynalon
Supplier: Dynalon
421125 421145 421135 421165 421115 421155
30620-370CS 313.51 USD InStock
30620-370 30617-164 30620-368 30620-366 30620-374 30620-372
Jar, Straight Sided, Polypropylene, Dynalon
Jars General Use Jars
Straight-sided polypropylene containers are supplied with white polypropylene screw closures
These autoclavable, translucent jars have many laboratory and industrial applications. Lightweight and easy to fill with a maximum operating temperature of 135°C (275°F).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty