Would you like to log in again?
Straight-Sided Container, Polystyrene, Dynalon
Supplier: Dynalon
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
421235
30620-376CS 278.44 USD InStock
30620-376
Straight-Sided Container, Polystyrene, Dynalon
Jars General Use Jars
Straight-sided jars are economical and ideal for storing lab specimens, supplies, or powders.
Straight-Sided Containers are supplied with white polypropylene screw closures. These “glass-clear” polystyrene jars have many uses in the lab. They are lightweight and easy to fill. Maximum temperature polystyrene is 70 °C.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty