Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Hydrometer Racks, Polypropylene, Dynalon
Supplier: Dynalon
103215-0001 103215-0000
30620-398EA 153.32 USD InStock
30620-398 30620-396
Hydrometer Racks, Polypropylene, Dynalon
Racks Hydrometer Racks
18-place polypropylene racks are specially designed to safeguard glass hydrometers
The three tiered rack with handles allows easy transport and storage of hydrometers. Top two plates have 1" diameter holes while the bottom plate has wells which taper from 1" to 3/8".
Ordering information: Additional sizes and materials are available upon request.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty