Would you like to log in again?
Stir Bar, Cross Head, PTFE, Dynalon
Supplier: Dynalon
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
304495-0003 304495-0001 304495-0004
30620-408CS 246.5 USD InStock
30620-408 30620-406 30620-410
Stir Bar, Cross Head, PTFE, Dynalon
Stirrers Magnetic Stirring Bars
Designed for round or flat bottom labware and test tubes
Greater mixing action at bottom, side, and top of container, even at low stirring speeds. Powerful permanent Alnico V magnetic cores. PTFE offers excellent chemical resistance and is autoclavable. Always store in pairs to maintain magnetic strength.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty