Would you like to log in again?
Stir Bar, Paddle-Shaped, PTFE, Dynalon
Supplier: Dynalon
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
304455-0002 304455-0003 304455-0004 304455-0005
30620-480CS 161.01 USD InStock
30620-480 30620-484 30620-474 30620-482
Stir Bar, Paddle-Shaped, PTFE, Dynalon
Stirrers Magnetic Stirring Bars
Stir bars are designed in a paddle configuration
Excellent mixing at slow speeds for biologiclal applications. Features larger vortex for more complete mixing. Powerful permanent Alnico V magnetic cores. PTFE offers excellent chemical resistance, and allow operation up to 260°C.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty