Would you like to log in again?
Stirring Rod, Solid, Dynalon
Supplier: Dynalon
303334-0003 303334-0004 303334-0000 303334-0001 303334-0002
30620-508CS 213.37 USD InStock
30620-508 30620-506 30620-504 30620-512 30620-510
Stirring Rod, Solid, Dynalon
Stirrers Stirring Rods
Pure PTFE solid stirring rod with flat tapered ends
Inert and will not scratch, the rod can be used up to 280°C. PTFE has excellent chemical resistance and is autoclavable.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty