Would you like to log in again?
PFA Vials, Savillex
Supplier: Savillex
200-030-20 200-003-20 200-007-30 200-015-30 200-007-20 200-022-20 200-015-20
30621-028EA 39.38 USD InStock
30621-028 30621-024 30621-034 30621-026 30621-036 30621-030 30621-032
PFA Vials, Savillex
Vials
Used for sample digestion and/or storage.
  • Service temperature range: –200 to 260 °C (–328 to 500 °F)
The flat exterior bottom surface and high service temperature range of these PFA vials make them ideal for use directly on a hot plate. Manufactured from the highest purity PFA resin, these vials are used for sample digestion prior to ultratrace metals analysis. Also ideal for a wide range of other uses. Available with round or conical shaped interior.
Ordering information: Closures are not included, and must be ordered separately. Vial trays include ID cards.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty