Would you like to log in again?
DST-1000 Acid Purification System, Savillex
Supplier: Savillex
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
525-1000-115
30621-038EA 20086.87 USD InStock
30621-038
DST-1000 Acid Purification System, Savillex
Water Purification Systems Stills
The Savillex DST-1000 Acid Purification System provides a cost effective and efficient supply of freshly distilled high-purity acid to your lab
Extremely easy to use, and requiring no cooling water, the DST-1000 can produce high purity (10ppt grade) HNO3, HCl, and HF acids from low cost trace metal (1ppb) grade. And since the acid is freshly produced on demand, it is always of the highest quality. Depending on your acid usage, the DST-1000 will pay for itself in months or even weeks.
Certifications: CE certified and RoHS compliant.
Ordering information: Includes 1L PFA collection bottle and stand.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty