Would you like to log in again?
Boston Round Glass Bottles, Narrow Mouth, Amber, C&G Containers
Supplier: C&G Containers
LAB004900000 LAB008100100 LAB016900000 LAB008100000 LAB033900000 LAB008900003 LAB033100110 LAB004469002 LAB033200000 LAB004200010 LAB033100000 LAB032260010 LAB032200012 LAB004100002 LAB008900000 LAB008100302 LAB004200110 LAB032100000 LAB008200010 LAB033200000B LAB016200000 LAB016200010 LAB032901000 LAB032900100 LAB032230010 LAB016100010 LAB033200010 LAB033290000 LAB032210010 LAB004100000 LAB033785000 LAB016100000
71000-594CS 75.42 USD InStock
71000-594 30621-138 71000-610 71000-578 71000-576 75839-246 10496-914 10496-916 71000-592 71000-590 76470-458 30621-144 30621-140 71000-608 71000-606 30621-142 71000-582 71000-600 71000-586 71000-620 75839-256 89107-152 71000-580 75839-252 30621-134 71000-614 30621-136 71000-612 71000-618 30621-130 30621-132 71000-616
Boston Round Glass Bottles, Narrow Mouth, Amber, C&G Containers
Bottles Glass Bottles
These glass jars are used primarily for the collection of liquid media, for testing semi-volatiles (light sensitive compounds), pesticides, PCBs, diesel range organics, TPH, metals, and phenolics.
Pre-cleaned and certified products contain the certificate of analysis and cleaning protocol form in the product case.
Pre-cleaned and certified products, a lot certificate of analysis is generated with the analytical results obtained including the concentration reporting limits and units of measurement. This certificate of analysis is included in each product case.
Wash protocols for these bottles include; A1 (Nitric acid wash), B1 (Deionized water wash), C1 (Hydrochloric acid wash), and L3 (no wash). Some bottles are also chemically preserved.
Secondary uses include containment and archiving. Please refer to the applicable Certificate of Analysis for certified analytes and corresponding reporting limits.
Ordering information: Container measurements and capacity 'contain' volumes may vary.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty