Would you like to log in again?
Ashless Quantitative Paper, Grade 74, Ahlstrom
Supplier: Ahlstrom
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
0740-1100 0740-1850 0740-0550 0740-0425 0740-3200 0740-2400 0740-0700 0740-0900 0740-4000 0074-0470 0740-12.50 0740-2700 0740-1500
47745-740PK 55.04 USD InStock
47745-740 47745-744 47745-742 47749-472 47749-470 47749-464 47749-476 47749-474 47749-468 47749-466 30621-278 38006-128 38006-106
Ashless Quantitative Paper, Grade 74, Ahlstrom
Filters Filter Paper Quantitative Filter Paper
Ahlstrom Grade 74 is a medium flow rate ashless (Ash 0.007%) filter paper suitable for routine quantitative gravimetric techniques.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty