Would you like to log in again?
SP Bel-Art 81-Place Polypropylene Freezer Storage Boxes, Bel-Art Products, a part of SP
Supplier: Bel-Art
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
F18852-0013 F18852-0011 F18852-0016
30621-404CS 839.99 USD InStock
30621-404 30621-398 30621-394
SP Bel-Art 81-Place Polypropylene Freezer Storage Boxes, Bel-Art Products, a part of SP
Boxes Cooling/Freezing Boxes and Bags
Polypropylene storage boxes provide compact storage and keep cryo vials organized on the bench, in the freezer, or during transport.
  • Polypropylene construction
  • Impervious to moisture and heavy use
  • Color-coded for quick visual identification
  • Alphanumeric grid for tracking individual samples
  • Does not fit a standard 2” freezer rack
Useful at temperatures –80°C to +121°C (-112°F to +249°F), these 81-place, 9 x 9 row boxes hold 0.5, 1.5 and 2.0 mL tubes and cryo vials
Five color choices are useful for organization and quick visual identification of samples. Secure fitting, lift-off, clear translucent lids let you easily view contents and have matte panels on two sides for trouble-free labeling. An alphanumeric grid lets you track individual samples. ‘No slide’ features molded in bottom and lid provide secure stacking.
Unlike cardboard models, these sturdy boxes will not break down with exposure to moisture and heavy use.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty