Would you like to log in again?
HiPrep SP HP Columns
Supplier: Cytiva
HiPrep™
29018183 29-0181-83
30621-418EA 1359.51 USD InStock
30621-418 CA30621-418
HiPrep SP HP Columns
Chromatography Columns
HiPrep SP HP 16/10 is a strong cation exchanger designed for preparative ion exchange chromatography separations. This 20 mL column is prepacked with SP Sepharose High Performance chromatography resin. The resin is based on rigid, highly cross-linked beaded agarose with a mean bead diameter of 34 µm for high resolution separations.
  • For high-performance preparative scale separations where high efficiency and high resolution are desired
  • Highly cross-linked agarose provides excellent chemical and physical stability
  • Slim, modern design allows for easy handling and connection
High mechanical strength of the matrix allows for high flow rates, together with outstanding resolution. Capacities vary according to the protein applied and its concentration. Loading capacity will also vary depending on the salt concentration, pH, and temperature at which the protein is applied.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty