Would you like to log in again?
Condensers, Reflux, Coil Type, Removable Hose Connections, Chemglass
Supplier: Chemglass
CG-1213-J-HC02 CG-1213-J-HC01
30622-516EA 785.07 USD InStock
30622-516 30622-518
Condensers, Reflux, Coil Type, Removable Hose Connections, Chemglass
Condensers
Coil style reflux condenser feature a larger diameter jacket.
  • Hose connections are on the bottom of the condenser
  • Makes it easier to connect hose/tubing connections are GL14 polypropylene hose barbs and screw caps
Coil style reflux condenser with a larger diameter jacket and coil, thereby providing maximum cooling capacity, while being shorter in length. GL hose connections provide a safe and easy way of connecting flexible tubing to a condenser. Jacket has a O.D. of 41 mm while the coil O.D. is 8 mm. Excellent for use with high vapor pressure solvents. Condenser has a top outer standard taper joint and lower inner drip tip joint.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty