Would you like to log in again?
Reflux Condensers, Double Cooling, Chemglass
Supplier: Chemglass
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
CG-1217-A-29 CG-1217-A-24
30622-522EA 593.74 USD InStock
30622-522 30622-520
Reflux Condensers, Double Cooling, Chemglass
Condensers
These high efficiency reflux condensers feature a double surface area with a cooling finger sealed into the secondary cooling jacket.
  • This condenser designs works best with closed loops chillers
Serrated hose connections for water connection extend through the outer bulb and have an O.D. of 10 mm at the middle serration.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty