Would you like to log in again?
Adapters, Distilling/Splitting, Process Reactors, Chemglass
Supplier: Chemglass
CG-1235-A-24 CG-1235-A-29
30622-524EA 752.08 USD InStock
30622-524 30622-526
Adapters, Distilling/Splitting, Process Reactors, Chemglass
Distillation Components Distillation Adapters
Compact and simple distilling/splitter adapter for use with process reactors.
  • Control valve is 0 to 4 mm PTFE
  • A 1/2" O.Ø distillate takeoff for use with compression fittings and TFE tubing
  • Plugs are supplied complete with Perfluoro O-rings
Vapor flows upward freely through the large vapor holes to the condenser. Distillate flow is directed using the valve to either the receiving flask or back to the reactor.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty