Would you like to log in again?
Chem-Vac™ Universal Stirrer Glass Ball Bearings, Chemglass
Supplier: Chemglass
CHEMVAC®
CG-2077-U-50 CG-2077-U-51 CG-2077-U-02 CG-2077-U-01
30622-656EA 949.2 USD InStock
30622-656 30622-654 30622-658 10034-432
Chem-Vac™ Universal Stirrer Glass Ball Bearings, Chemglass
Stirrers Stirring Tools
These universal Chem-Vac™ PTFE stirrer bearings provides a vacuum tight flake-free seal on PTFE, glass or stainless-steel stir shafts. Loosening nut above the standard taper inner joint allows easy and quick removal.
  • Flake-free
  • Chemically resistant
  • The polymer blend sleeve provides a long-lasting seal for use up to 500 rpm
  • For use with polished glass, PTFE or stainless-steel stirrer shafts
The upper portion of bearing rotates with the stirrer shaft providing a flake-free seal. The bearing consists of a PTFE standard taper body, glass filled polypropylene screw cap, PEEK®/PTFE sleeve and a glass filled polypropylene loosening nut and a specially designed glass ball-bearing for rigidity and smoothness of operation.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty