Would you like to log in again?
Media Bottles, PETG, Square, Chemglass
Supplier: Chemglass
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
CLS-1150-0125 CLS-1150-2000 CLS-1150-0500 CLS-1150-0250 CLS-1150-0060 CLS-1150-0030 CLS-1150-1000
30622-700PK 95.26 USD InStock
30622-700 30622-710 30622-704 30622-702 30622-712 30622-708 30622-706
Media Bottles, PETG, Square, Chemglass
Bottles Cell Culture Bottles
Break-resistant and durable media bottles are manufactured from polyethylene terephthalate copolyester (PETG) and graduated 'to contain'.
  • Transparent, optically clear, and leak-proof
  • Freezable at –20 °C
  • Gamma Irradiated and Sterility Assurance Level (SAL) 10⁻⁶
  • Pyrogen, DNase, RNase and particle-free bottles
Bottles are square with rounded edges to insure an 'Easy-Grip' for pouring. Cap is manufactured with polyethylene (HDPE) and comes with a security ring which provides a tamper-evident seal. The cap is liner-free and comes with a leak-proof guarantee. Bottles are inexpensive alternatives to glass media bottles, and provide reduced permeability to both CO₂ and O₂. Bottles are radiation sterilized and non-pyrogenic.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty