Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Autoclave Bags, Chemglass
Supplier: Chemglass
CLS-1192-2430 CLS-1192-2436 CLS-1192-1224
30622-742CS 240.51 USD InStock
30622-742 30622-738 30622-740
Autoclave Bags, Chemglass
Bags Sterilization Pouches
These durable 0.002'' thick transparent polypropylene autoclave bags provides maximum protection and performance for autoclavability.
  • These bags can hold 65, 200 and 240 standard size petri dishes
  • Steam autoclavable at 135 °C
It is used to sterlize labware such as Petri dishes, membrane filters, culture plates, pipettes, and more.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty