Would you like to log in again?
Spectrophotometer Cuvettes, Chemglass
Supplier: Chemglass
CLS-1375-007 CLS-1375-105 CLS-1375-103 CLS-1375-101 CLS-1375-003 CLS-1375-005 CLS-1375-107 CLS-1375-001
30622-766PK 47.56 USD InStock
30622-766 30622-754 30622-764 30622-758 30622-768 30622-756 30622-762 30622-760
Spectrophotometer Cuvettes, Chemglass
Cuvettes Spectrophotometer Cuvettes
Disposable cuvettes, made of polystyrene (PS) or polymethyl methacrylate (PMMA), are an excellent choice for most spectrophotometry applications in the visible range.
  • Provide accurate, reliable results, even at wavelengths as low as 300 nm
  • Macro and semi-micro sizes to accommodate most sample volumes
  • Include an arrow mark to indicate direction of transmission and reduce variation
  • Compatible with most spectrophotometers and photometers
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty