Would you like to log in again?
Delong Shaker Flasks with Three Extra Deep Side Baffles, Chemglass
Supplier: Chemglass
CLS-2046-04 CLS-2046-10 CLS-2046-05 CLS-2046-03 CLS-2046-01 CLS-2046-06 CLS-2046-08 CLS-2046-02
30623-234EA 79.13 USD InStock
30623-234 30623-248 30623-236 30623-238 76380-146 30623-240 30623-242 30623-244
Delong Shaker Flasks with Three Extra Deep Side Baffles, Chemglass
Flasks Erlenmeyer Flasks
Extra deep baffle indents on the lower outside edge of the flask to maximize oxygen transfer to the culture medium when used on an orbital shaker.
  • Shaker flasks are made of Class A borosilicate glass Type 1
Delong style neck is made to fit stainless steel or plastic closures, while the longer neck reduces the amount of splashing that would reach the closure.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty