Would you like to log in again?
Sealing Films, Sealplate, Chemglass
Supplier: Chemglass
CLS-4001-001 CLS-4001-003
30623-766PK 97.6 USD InStock
30623-766 30623-768
Sealing Films, Sealplate, Chemglass
Seals General Purpose Sealing Films
Polyester sealing films minimize evaporation, prevent spillage and contamination between wells, and provide a secure seal. Secure sealing of all wells eliminates edge effects in sensitive ELISA assays.
  • Functional temperature range from –40 to 120 °C
  • Available sterile or non-sterile
  • Sealing films are non-pierceable
SealPlate® sealing films can also be used with tissue culture plates for short-term storage, incubation, and containment of biohazards.
Each SealPlate® sealing film measures 79.4×146.1 mm.
Packaging: Packed in boxes of 100.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty