Would you like to log in again?
Sealing Films, Aeraseal 96, Chemglass
Supplier: Chemglass
CLS-4001-013 CLS-4001-015
30623-784PK 140.64 USD InStock
30623-784 30623-786
Sealing Films, Aeraseal 96, Chemglass
Seals General Purpose Sealing Films
A 114 µm thick hydrophobic porous sealing film with medical-grade adhesive for tissue culture plates, bio-blocks, and 96-well plates where gas exchange is necessary for cell or bacterial growth
  • Non-Cytotoxic, Highly Gas Permeable
  • Easily Pierceable with Pipet Tips for Sample Recovery
  • Recommended for Temperatures from -20° to +80°C
AeraSeal™ sealing films minimize cross-contamination, spillage and evaporation. They allow uniform air and CO2 exchange for all wells, unlike plate lids which favor exchange for the wells near the plate edges. Dimensions: 82.6 x 142.9 mm for standard-size tissue culture plates.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty