Would you like to log in again?
Double-Sided Tape, Chem-glass
Supplier: Chemglass
CLS-4008-T03
30623-822EA 21.84 USD InStock
30623-822
Double-Sided Tape, Chem-glass
Tapes
This single-use, double-sided adhesive tape is designed for use with rockers and shakers.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty