Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Dura-Mag™ Magnetic Stirrers, 5-position, Chemglass
Supplier: Chemglass
CLS-4100-04 CLS-4100-A4 CLS-4100-06 CLS-4100-A6
30624-012EA 4560.93 USD InStock
30624-012 30624-010 30624-020 30624-018
Dura-Mag™ Magnetic Stirrers, 5-position, Chemglass
Stirrers Magnetic Hotplate Stirrers, Multi-position
The five position magnetic stirrer is a rugged design for 24/7 laboratory or production applications. It is ideal for CO2 incubators with a maximum capacity of five 1000 ml spinner vessels.
  • Trouble free brushless motor
  • Heavy duty magnets and drive system
  • Stainless steel top with easy alignment marks
  • Non-slip trouble free toothed belt design for long life
  • Illuminated LCD display
The factory installed alarm has 85 dB internal audible buzzer, serial port for external alarm configuration (external alarm is not supplied), convenient alarm reset button and the alarm state is initiated if any of the magnets stops spinning.
Ordering information: Optional output for remote communication and motion alarm available.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty