Would you like to log in again?
Air Jet Test Sieve, W.S. Tyler™
Supplier: W.S. Tyler
9410 9003
30624-364EA 619.84 USD InStock
30624-364 30624-362
Air Jet Test Sieve, W.S. Tyler™
Sieves Test Sieves
For use with air jet sieving systems.
  • Overall Height: 4.44 cm (13/4")
  • Depth to Cloth: 2.86 cm (11/8")
Each sieve is serialized and supplied with a “Test Sieve Certificate.” Sieves are marked with the USA Sieve Number.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty