Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
Chemware® Teflon® FEP Gas Sampling Bags, Saint-Gobain Performance Plastics
Supplier: Saint-Gobain
Chemware®
D1075003-10 D1075007-10 D1075005-10 D1075001-10 D1075002-10
32310-300CS 999.87 USD InStock
32310-300 32310-302 32310-301 32310-304 32310-306
Chemware® Teflon® FEP Gas Sampling Bags, Saint-Gobain Performance Plastics
Bags Gas Bags
For gas sample analysis, sampling smokestack exhaust systems, monitoring air quality, and calibrating instruments in the field.
  • Inert to almost all chemicals
  • Low permeability limits sample loss
  • Pressure less, and fill is rated by volume
Made from 5 mil thick FEP fluoropolymer film, these flexible, sturdy bags are heat-sealed on all sides for complete chemical resistance and leakproof operation. Available with either a nickel-plated brass on/off valve which accepts ¹/₄" Int.Ø. flexible tubing or PTFE/silicone septum in polypropylene housing, for use with a hypodermic needle.
Packaging: Individually packaged.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty