Would you like to log in again?
Falcon® Individually Wrapped Serological Pipettes, Corning
Supplier: Corning
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Falcon®
356543 357521 357525 356551 357551 357600 357550 356550 357525 357535 357507 356551 356507 357543 356521 356525 356521 357535 356507 357550 356535 356525 356543 357507 356535 357543 357521 357600 357551 356550
32314-008CS 180.07 USD InStock
CA12777-018 32314-008 32314-006 CA53300-283 CA53106-441 32314-012 32314-010 CA32314-002 CA32314-000 53106-195 CA53300-421 CA53300-363 12777-018 CA53300-567 CA53300-523 CA32314-006 CA32314-004 CA32314-008 32314-000 CA32314-010 53106-441 53300-283 CA53106-195 32314-004 32314-002 CA32314-012 53300-567 53300-523 53300-421 53300-363
Falcon® Individually Wrapped Serological Pipettes, Corning
Pipettes Serological Pipettes
The pipettes graduation alignment system is a fast and simple way to bring graduations to your line of sight. Polystyrene, disposable pipettes for tissue culture, bacteriological, and research applications.
  • Sterile
  • RNase-/DNase-free
  • Noncytotoxic, nonpyrogenicity tested to less than 0.1 EU/ml
  • Color-coded package and markings for ease of identification
Simply pop open or peel back the top of the pipette package and line up the pipettor before completely removing the pipette from its wrapping. When you remove the pipette from its package, the graduations should face you every time.
Packaging: Individually wrapped in plastic-peelable bags, recyclable.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty