Would you like to log in again?
OxiClear™ Disposable Inline Gas Purifiers
Supplier: Diamond Tool & Die
OxiClear™
DGP-125-R2 DGP-125-R1 DGP-250-R2 DGP-250-R1
32574-076EA 549.76 USD InStock
32574-076 32574-054 32574-065 32574-021
OxiClear™ Disposable Inline Gas Purifiers
Filters Gas Filters
Ideal for gas chromatography, glove boxes, and anaerobic experiments.
Oxygen-absorbing gas purifiers with high efficiency and low resistance to gas flow. Reduce amount of oxygen, trace amounts of water, and organic compounds from inert carrier gases such as nitrogen, helium, argon, and hydrogen. Tubular inline configuration with nickel-plated brass, threaded, male tube compression fitting on both ends.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty