Would you like to log in again?
Dual-Compartment Dispenser, AK®
Supplier: AK
AK-1478
32799-016EA 538.81 USD InStock
32799-016
Dual-Compartment Dispenser, AK®
PPE Dispensers
Designed to keep an assortment of disposable products in one location, this large-capacity dispenser features two compartments
All-purpose dispenser is made of clear 6 mm (¹/₄") acrylic and is ideal for shoe covers, gloves, bouffants, hairnets, beard covers, and hoods. Openings feature radius edges for a smooth finish.
Compartment dimensions: 24.1× 30.5×45.7 cm (9.5×2×18")
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty