Would you like to log in again?
GLOVE LINER DISPENSER CLR 11X7X11
Supplier: APPLIED PLASTICS MACHINING, INC
Glove Liner with 4 inch Round Hole and Lid^ Easy loading and retrieval of glove liners. Wall mount or free standing. Made of 3/16 in. clear acrylic. 11 in. W x 7 in. H x 11 in. D.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
GLOVE LINER DISPENSER CLR 11X7X11 32799-054 AK-787 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty