Would you like to log in again?
Flip-Top Finger Cot Dispenser, AK®
Supplier: AK
AK-901
32799-056EA 146.36 USD InStock
32799-056
Flip-Top Finger Cot Dispenser, AK®
Glove Dispensers and Glove Box Holders
Cleanroom finger cot dispenser features hinged lid and door for easy loading and removal.
  • Made of clear 5 mm (3/16") acrylic
  • Openings feature radius edges for a smooth finish
Dispenser can be placed on a countertop or mounted on a wall.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty