Would you like to log in again?
Four-Edged Glass Scoring Knife, Wale Apparatus
Supplier: Wale Apparatus
90-6050
32861-011EA 336.03 USD InStock
32861-011
Four-Edged Glass Scoring Knife, Wale Apparatus
Knives
The four cutting edges of this knife ensures a greater ease of use and longer life and it is ideally designed for cutting glass and quartz tubing and rods.
  • Retractable and replaceable blade
Unique square collet holds high-quality tungsten-carbide steel blade. User can expose as much of blade as desired and easily replace a worn blade.
Ordering information: Knife comes complete with four-edged blade. Replacement blades available.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty