Would you like to log in again?
GLOVES CRYO WP WRIST MD PK=1PR
Supplier: TEMPSHIELD INC
Cryo-Gloves*^ (Wrmwp) For Protection In Ultra-Cold (-160[Degree]C) To High-Heat (150[Degree]C). Use With Liquid Nitrogen Vapor Phase, Blast Freezers, Autoclaves And Sterilizers. Solvent Resistant. Not For Immersion In Liquid Nitrogen. 100% Waterproof. Wrist Length, 30.5Cm Long. Size: Medium.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
GLOVES CRYO WP WRIST MD PK=1PR 32884-022 WRMWP Case of 20 Retrieving
Pack of 2 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty