Butyl gloves exhibit high permeation resistance to gas and water vapors.
- High permeation resistance to gas and water vapors
- Resistant to oxygenated solvents and most oxidizing chemicals
- Remain flexible and sensitive, even at lower temperatures
- Hand specific, curved design improves fit and comfort
- Rolled cuff
Gloves stay flexible and sensitive, even at lower temperatures. Curved finger and hand design provide a comfortable fit. Gloves with a rough finish are excellent for wet applications. Rolled cuff. Hand-specific.
Resistant to oxygenated solvents and most oxidizing chemicals.