Would you like to log in again?
Chemsoft™ CE Nitrile Gloves, Honeywell Safety
Supplier: Honeywell Safety
Chemsoft™
CE412W/XL CE412W/M CE412W/S CE412W/L CE412W/XS
32888-028CS 760.44 USD InStock
32888-028 32888-026 32888-020 32888-024 32888-022
Chemsoft™ CE Nitrile Gloves, Honeywell Safety
Gloves
Strong, supple 100% nitrile gloves for the cleanroom offer good chemical resistance and superior tactility.
  • Double bagged to ensure cleanliness
  • Multiple washings assure low particulate and extractable levels
  • Textured fingertips provide a good grip
  • Ambidextrous and powder-free
  • Antistatic
Rolled cuff prevents tearing while donning. Do not contain allergy-causing latex proteins. Color: white.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty