Would you like to log in again?
Nitrile Gloves, Honeywell Safety
Supplier: Honeywell Safety
Nitty-Gritty®
LA102G/9 LA102G/11 LA172G/9-H5 LA102G/11 LA132G/8 LA132G/11 LA172G/10 LA132G/10-H5 LA132G/9 LA132G/8 LA102G/10 LA102G/9 LA172G/8 LA132G/7 LA102G/7 LA102G/10 LA172G/7 LA172G/8 LA132G/11 LA102G/7 LA102G/8 LA132G/9 LA102G/8
32888-274CS 475.41 USD InStock
32888-274 CA32888-240 32888-284 32888-240 CA32888-274 32888-272 CA32888-242 32888-282 32888-278 CA32888-280 32888-244 CA32888-270 32888-276 CA32888-282 CA32888-272 32888-286 32888-242 CA32888-248 32888-248 32888-246 CA32888-244 CA32888-278 CA32888-246
Nitrile Gloves, Honeywell Safety
Gloves
Anatomically shaped, flexible gloves feature a straight cuff edge for good tear resistance.
  • Styles with flocked interior give the glove a soft, pleasant feel and absorb perspiration
  • Case hardened for easy donning and added toughness
High nitrile buna rubber (NBR) content provides superior resistance to cuts, snags, abrasion, and punctures. Available in many thicknesses to meet your needs.
Certifications: Comply with USDA and FDA regulations, 21CFR, for use in food processing.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty