Would you like to log in again?
Magid® CleanMaster® Nylon Tricot Gloves, Magid
Supplier: Magid
7402XL 7402L 7402M
32889-312CS 2535.46 USD InStock
32889-312 32889-308 32889-310
Magid® CleanMaster® Nylon Tricot Gloves, Magid
Gloves
These gloves are comfortable and durable for cleanroom, paint and electronics projects.
  • Two-piece nylon tricot glove
  • Low linting for less contamination
  • Color-coded hem for easy matching
This style is hemmed, with a two-piece cut and sewn pattern. These gloves are reversible and fits either hand, ensuring longer wear and greater ease in sorting after laundry.
An insert thumb provides a better fit, giving this comfortable style the wear of a dress glove.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty